missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,4,4,4-Heptafluorobutyraldehyde hydrate, tech.

Catalog No. AAL0087514
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0087514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0087514 Supplier Thermo Scientific Chemicals Supplier No. L0087514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 375-02-0 | C4HF7O | 198.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 375-02-0
Molecular Formula C4HF7O
Molecular Weight (g/mol) 198.04
MDL Number MFCD00039731
InChI Key IQJZGNJYXIIMGP-UHFFFAOYSA-N
Synonym heptafluorobutyraldehyde, butanal, heptafluoro, butyraldehyde, heptafluoro, heptafluorobutyraldehyde, tech., perfluorobutanal, perfluorobutyraldehyde, c3f7cho, heptafluoro-n-butyraldehyde, butanal,2,2,3,3,4,4,4-heptafluoro
PubChem CID 67809
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutanal
SMILES FC(F)(F)C(F)(F)C(F)(F)C=O

Specifications

Refractive Index 1.32
Quantity 25 g
Beilstein 1367365
Formula Weight 198.04 (Anhydrous)
Grade Technical
Chemical Name or Material 2,2,3,3,4,4,4-Heptafluorobutyraldehyde hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-783-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only