missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,4,4,4-Heptafluoro-1-butanol, 95%

Catalog No. AC243890500
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC243890500 50 g Glass bottle
AC243890100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC243890500 Supplier Thermo Scientific Chemicals Supplier No. 243890500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 375-01-9 | C4H3F7O | 200.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 375-01-9
Molecular Formula C4H3F7O
Molecular Weight (g/mol) 200.06
MDL Number MFCD00004674
InChI Key WXJFKAZDSQLPBX-UHFFFAOYSA-N
Synonym 2,2,3,3,4,4,4-heptafluoro-1-butanol, 1-butanol, 2,2,3,3,4,4,4-heptafluoro, 1h,1h-heptafluorobutanol-1, 1h,1h-heptafluoro-1-butanol, 1,1-dihydroperfluorobutyl alcohol, 2,2,3,3,4,4,4-heptafluorobutanol, 1,1-dihydroperfluorobutanol, 1,1-h,h-heptafluorobutanol, perfluoro-1,1-dihydrobutanol, butanol, 2,2,3,3,4,4,4-heptafluoro
PubChem CID 9776
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutan-1-ol
SMILES OCC(F)(F)C(F)(F)C(F)(F)F

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color White to Beige
Density 1.6000g/mL
Boiling Point 96°C to 97°C
Flash Point 25°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.2920 to 1.294
Linear Formula CF3CF2CF2CH2OH
Quantity 50 g
Beilstein 01, 1547
Specific Gravity 1.6
Solubility Information Solubility in water: slightly soluble
Formula Weight 200.06
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 2,2,3,3,4,4,4-Heptafluoro-1-butanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea
EINECSNumber 206-782-1

RUO – Research Use Only