missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethylamine hydrochloride, 98%

Numéro de catalogue. p-7022229
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAA1490409 10 g
AAA1490418 50 g
AAA1490422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AAA1490409 Fournisseur Thermo Scientific Chemicals Code fournisseur A1490409
Il en reste null

CAS: 373-88-6 | C2H5ClF3N | 135.51 g/mol

2,2,2-Trifluoroethylamine hydrochloride, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,2-Trifluoroethylamine hydrochloride, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff

Solubility
It is Soluble in 0.7 to water, but soluble in ethanol and chloroform, slightly soluble in benzene, hardly soluble in ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic. Keep away from strong oxidizing agents, acids, acid chlorides, acid anhydrides.

Identifiants chimiques

CAS 373-88-6
Molecular Formula C2H5ClF3N
Molecular Weight (g/mol) 135.51
MDL Number MFCD00012875
InChI Key ZTUJDPKOHPKRMO-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethylamine hydrochloride, 2,2,2-trifluoroethanamine hydrochloride, trifluoroethylamine hydrochloride, ethanamine, 2,2,2-trifluoro-, hydrochloride, 2,2,2-trifluoroethylaminehydrochloride, 2,2,2-trifluoroethylamine, hydrochloride, 2,2,2-trifluoroethan-1-amine hydrochloride, 2,2,2-trifluoroethylamine hcl, ethylamine, trifluoro-, hydrochloride, ethanamine, 2,2,2-trifluoro-, hydrochloride 1:1
PubChem CID 9772
IUPAC Name 2,2,2-trifluoroethanamine;hydrochloride
SMILES [H+].[Cl-].NCC(F)(F)F

Spécifications

Melting Point 218°C (sublimation)
Linear Formula CF3CH2NH2·HCl
Quantity 10 g
Beilstein 3652103
Sensitivity Hygroscopic
Solubility Information It is Soluble in 0.7 to water,but soluble in ethanol and chloroform,slightly soluble in benzene,hardly soluble in ether.
Formula Weight 135.52
Percent Purity 98%
Chemical Name or Material 2,2,2-Trifluoroethylamine hydrochloride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 206-771-1
RTECSNumber KS0250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only