missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethyl trifluoroacetate, 97+%, Thermo Scientific Chemicals

Catalog No. AAA1828314
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1828314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1828314 Supplier Thermo Scientific Chemicals Supplier No. A1828314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 407-38-5 | Molecular Formula: C4H2F6O2 | Molecular Weight: 196.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 407-38-5
Molecular Formula C4H2F6O2
Molecular Weight (g/mol) 196.05
MDL Number MFCD00000418
InChI Key ZKUJOCJJXCPCFS-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethyl trifluoroacetate, acetic acid, trifluoro-, 2,2,2-trifluoroethyl ester, trifluoroacetic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyltrifluoroacetate, pubchem12623, acmc-1ake7, cf3co2ch2cf3, cf3c o och2cf3, ksc496e0r, trifluoroethyl trifluoroacetate
PubChem CID 67888
IUPAC Name 2,2,2-trifluoroethyl 2,2,2-trifluoroacetate
SMILES FC(F)(F)COC(=O)C(F)(F)F

Specifications

Melting Point -65°C
Density 1.464
Boiling Point 54°C to 55°C
Flash Point 0°C (32°F)
Refractive Index 1.277
Quantity 25 g
UN Number UN2924
Beilstein 1783427
Sensitivity Moisture sensitive
Formula Weight 196.05
Percent Purity 98%
Chemical Name or Material 2,2,2-Trifluoroethyl trifluoroacetate
Show More Show Less
Hazard Category H225-H314
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 206-985-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only