missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethyl p-toluenesulfonate, 98+%

Catalog No. AAA1787122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1787122 100 g
AAA1787114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1787122 Supplier Thermo Scientific Chemicals Supplier No. A1787122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 433-06-7 | C9H9F3O3S | 254.223 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 433-06-7
Molecular Formula C9H9F3O3S
Molecular Weight (g/mol) 254.223
MDL Number MFCD00000443
InChI Key IGKCQDUYZULGBM-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethyl p-toluenesulfonate, 2,2,2-trifluoroethyl tosylate, trifluoroethanol tosylate, trifluorethyltosylate, 2,2,2-trifluoroethyl-p-toluenesulfonate, ethanol, 2,2,2-trifluoro-, 4-methylbenzenesulfonate, 2,2,2-trifluoroethyl p-toluenesulphonate, ethanol, 2,2,2-trifluoro-, p-toluenesulfonate, 2,2,2-trifluoroethyl 4-methylbenzene-1-sulfonate, ethanol, 2,2,2-trifluoro-, 1-4-methylbenzenesulfonate
PubChem CID 67942
IUPAC Name 2,2,2-trifluoroethyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC(F)(F)F

Specifications

Melting Point 37°C to 42°C
Boiling Point 87°C to 92°C (0.1 mmHg)
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 2699394
Sensitivity Moisture sensitive
Formula Weight 254.23
Percent Purity ≥98%
Chemical Name or Material 2,2,2-Trifluoroethyl p-toluenesulfonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-085-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only