missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethyl methacrylate, 99%, stabilized, Thermo Scientific Chemicals

Catalog No. AC312780250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC312780250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC312780250 Supplier Thermo Scientific Chemicals Supplier No. 312780250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 352-87-4
Molecular Formula C6H7F3O2
Molecular Weight (g/mol) 168.12
MDL Number MFCD00013576,MFCD00274739
InChI Key QTKPMCIBUROOGY-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethyl methacrylate, trifluoroethyl methacrylate, methacrylic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethylmethacrylate, 2-propenoic acid, 2-methyl-, 2,2,2-trifluoroethyl ester, 2-methyl-acrylic acid 2,2,2-trifluoro-ethyl ester, 2-propenoic acid, 2-methyl-, trifluoroethyl ester, 2-methyl-2-propenoic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyl m, acmc-1af8z
PubChem CID 9608
IUPAC Name 2,2,2-trifluoroethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC(F)(F)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color White
Density 1.1800g/mL
Boiling Point 59.0°C (100.0 mmHg)
Flash Point 16°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3600 to 1.3620
Linear Formula H2C=C(CH3)CO2CH2CF3
Quantity 25 g
Specific Gravity 1.18
Stabilizer 100ppm MEHQ
Formula Weight 168.12
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2,2,2-Trifluoroethyl methacrylate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-525-3
TSCA TSCA

RUO – Research Use Only