missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethanesulfonyl chloride, 95%

Catalog No. AC305350100
Change view
Click to view available options
Quantity:
1 g
2.5 g
10 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC305350100 10 g Glass bottle
AC305350010 1 g Glass bottle
AC305350025 2.5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC305350100 Supplier Thermo Scientific Chemicals Supplier No. 305350100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1648-99-3 | C2H2ClF3O2S | 182.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1648-99-3
Molecular Formula C2H2ClF3O2S
Molecular Weight (g/mol) 182.55
MDL Number MFCD00007458
InChI Key CXCHEKCRJQRVNG-UHFFFAOYSA-N
Synonym tresyl chloride, 2,2,2-trifluoroethane-1-sulfonyl chloride, ethanesulfonyl chloride, 2,2,2-trifluoro, 2,2,2-trifluoroethanesulphonyl chloride, 2,2,2-trifluoroethyl sulphonyl chloride, 2,2,2-trifluoroethylsulfonyl chloride, chloro 2,2,2-trifluoroethyl sulfone, 2,2,2-trifluoroethanesulfonylchloride, ethanesulfonylchloride, 2,2,2-trifluoro, pubchem2553
PubChem CID 74242
IUPAC Name 2,2,2-trifluoroethanesulfonyl chloride
SMILES C(C(F)(F)F)S(=O)(=O)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 1.6500g/mL
Boiling Point 64°C to 67°C (45.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Refractive Index 1.3870 to 1.389
Linear Formula CF3CH2SO2Cl
Quantity 10 g
Specific Gravity 1.65
Formula Weight 182.55
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 2, 2-Trifluoroethanesulfonyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-713-7

RUO – Research Use Only