missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroacetophenone, 98%

Catalog No. AAA1140322
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1140322 100 g
AAA1140306 5 g
AAA1140314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1140322 Supplier Thermo Scientific Chemicals Supplier No. A1140322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 434-45-7 | C8H5F3O | 174.122 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 434-45-7
Molecular Formula C8H5F3O
Molecular Weight (g/mol) 174.122
MDL Number MFCD00000420
InChI Key KZJRKRQSDZGHEC-UHFFFAOYSA-N
Synonym trifluoroacetophenone, 2,2,2-trifluoroacetophenone, 2,2,2-trifluoro-1-phenylethan-1-one, phenyl trifluoromethyl ketone, trifluoromethyl phenyl ketone, alpha,alpha,alpha-trifluoroacetophenone, ethanone, 2,2,2-trifluoro-1-phenyl, 2,2,2-trifluoro-1-phenyl-ethanone, trifluoroacetylbenzene, unii-6t7l1upy09
PubChem CID 9905
IUPAC Name 2,2,2-trifluoro-1-phenylethanone
SMILES C1=CC=C(C=C1)C(=O)C(F)(F)F

Specifications

Melting Point -40°C
Density 1.24
Boiling Point 165°C to 166°C
Flash Point 42°C (108°F)
Refractive Index 1.4595
Quantity 100 g
UN Number UN1224
Beilstein 1866286
Formula Weight 174.12
Percent Purity 98%
Chemical Name or Material 2,2,2-Trifluoroacetophenone
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 207-103-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only