missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trichloroethyl chloroformate, 97%, Thermo Scientific™

Numéro de catalogue. AC161810250
Modifier l'affichage
Click to view available options
25g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue.
AC161810250 25g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. AC161810250 Fournisseur Thermo Scientific Chemicals Code fournisseur 161810250

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Coupling reagent

Spécifications

Nom ou substance chimique 2, 2, 2-Trichloroethyl chloroformate
Nom Remarque 96%
CAS 17341-93-4
CAS % minimum 95.0
CAS % maximal 100.0
Densité 1.5390g/mL
Point d’ébullition 171°C to 172°C
Point d’éclair >100°C
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 96% min. (GC)
Emballage Glass bottle
Formule moléculaire C3H2Cl4O2
Indice de réfraction 1.4690 to 1.471
Formule linéaire ClCO2CH2CCl3
Quantité 25g
Numéro MDL MFCD00000810
Fieser 02,426; 03,296; 04,522; 05,686; 07,383
Gravité spécifique 1.539
Synonyme 2,2,2-trichloroethyl chloroformate, trichloroethyl chloroformate, 2,2,2-trichloroethylchloroformate, carbonochloridic acid, 2,2,2-trichloroethyl ester, 2,2,2-trichloroethoxycarbonyl chloride, chloroformic acid 2,2,2-trichloroethyl ester, formic acid, chloro-, 2,2,2-trichloroethyl ester, chloro 2,2,2-trichloroethoxy methanone, beta, beta, beta-trichloroethoxycarbonyl chloride, troc
Renseignements sur la solubilité Solubility in water: decomposes. Other solubilities: soluble in most common organic solvents
Clé InChI LJCZNYWLQZZIOS-UHFFFAOYSA-N
SOURIRES C(C(Cl)(Cl)Cl)OC(=O)Cl
Nom de l’IUPAC 2,2,2-trichloroethyl carbonochloridate
Poids moléculaire (g/mol) 211.86
PubChem CID 87063
Poids de formule 211.86
Pourcentage de pureté 97%
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 241-363-7
TSCA TSCA

RUO – Research Use Only