missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Tribromoethanol 98.0+%, TCI America™

Catalog No. T142025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T142025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T142025G Supplier TCI America Supplier No. T142025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75-80-9
Molecular Formula C2H3Br3O
Molecular Weight (g/mol) 282.757
MDL Number MFCD00004671
InChI Key YFDSDPIBEUFTMI-UHFFFAOYSA-N
Synonym tribromoethanol, avertin, bromethol, ethobrome, tribromethanol, ethobrom, basibrol, narcolan, narcotyl, narkolan
PubChem CID 6400
IUPAC Name 2,2,2-tribromoethanol
SMILES C(C(Br)(Br)Br)O

Specifications

Melting Point 81°C
Color White-Yellow
Boiling Point 94°C
Quantity 25 g
Formula Weight 282.76
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2,2-Tribromoethanol
EINECSNumber (2)-2002
RTECSNumber KM3675000
TSCA Yes