missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 2,2,2-Tribromoethanol

Numéro de catalogue. T4840225G
Encompass
Modifier l'affichage
Click to view available options
Quantité:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
T4840225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. T4840225G Fournisseur Sigma Aldrich Code fournisseur T4840225G
Il en reste null

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,2,2-Tribromoethanol

Spécifications

CAS 75-80-9
Point de fusion 73°C to 79°C (lit.)
Point d’ébullition 92°C to 93°C (10 mmHg)
Formule moléculaire C2H3Br3O
Formule linéaire Br3CCH2OH
Numéro MDL MFCD00004671
Quantité 25 g
Synonyme Tribromoethyl alcohol
Poids moléculaire (g/mol) 282.76
Pourcentage de pureté 97%