Learn More
2-[(11bR)-3H-Binaphtho[2,1-c:1',2'-e]phosphepin-4(5H)-yl]ethylamine, 97+%, Thermo Scientific Chemicals
Description
2-[(11bR)-3H-Binaphtho[2,1-c:1',2'-e]phosphepin-4(5H)-yl]ethylamine, is used as a application of innovative catalyst, ligand synthesis and application in asymmetric catalysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-[(11bR)-3H-Binaphtho[2,1-c:1′,2′-e]phosphepin-4(5H)-yl]ethylamine, is used as a application of innovative catalyst, ligand synthesis and application in asymmetric catalysis.
Solubility
Soluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Air, sensitive.
Specifications
Specifications
| Quantity | 250 mg |
| Sensitivity | Air Sensitive |
| Solubility Information | Soluble in water. |
| Formula Weight | 355.41 |
| Percent Purity | ≥97% |
| Chemical Name or Material | 2-[(11bR)-3H-Binaphtho[2,1-c:1',2'-e]phosphepin-4(5H)-yl]ethylamine |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only