missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Naphthyl)-5-phenyloxazole 99.0+%, TCI America™

Catalog No. N00681G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N00681G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N00681G Supplier TCI America Supplier No. N00681G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 846-63-9
Molecular Formula C19H13NO
Molecular Weight (g/mol) 271.319
MDL Number MFCD00019714
InChI Key WWVFJJKBBZXWFV-UHFFFAOYSA-N
Synonym 2-1-naphthyl-5-phenyloxazole, alpha-npo, oxazole, 2-1-naphthalenyl-5-phenyl, 2-1-naphthyl-5-phenyl-1,3-oxazole, 2-naphthalen-1-yl-5-phenyloxazole, oxazole, 2-1-naphthyl-5-phenyl, .alpha.-npo, anpo, 2-alpha-naphthyl-5-phenyloxazole, 2-naphthalen-1-yl-5-phenyl-1,3-oxazole
PubChem CID 70058
IUPAC Name 2-naphthalen-1-yl-5-phenyl-1,3-oxazole
SMILES C1=CC=C(C=C1)C2=CN=C(O2)C3=CC=CC4=CC=CC=C43

Specifications

Melting Point 106°C
Quantity 1 g
Formula Weight 271.32
Percent Purity ≥99.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-(1-Naphthyl)-5-phenyloxazole [for scintillation spectrometry]