missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Hydroxyethyl)-6-methoxynaphthalene 98.0+%, TCI America™

Catalog No. H12705G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H12705G Supplier TCI America Supplier No. H12705G

Chemical Identifiers

CAS 77301-42-9
Molecular Formula C13H14O2
Molecular Weight (g/mol) 202.25
MDL Number MFCD01632581
InChI Key OUVJWFRUESFCCY-SECBINFHSA-N
Synonym 1-6-methoxy-2-naphthyl ethanol, 1-6-methoxynaphthalen-2-yl ethanol, 1-6'-methoxy-2-naphthyl ethanol, +/--1-6-methoxynaphthyl ethanol, 2-1-hydroxyethyl-6-methoxynaphthalene, 1-6-methoxynaphthalen-2-yl ethan-1-ol, 6-methoxy-alpha-methyl-2-naphthalenemethanol, 1rs-1-6-methoxy-2-naphthyl ethanol naproxen impurity k, 1-6-methoxy-2-naphthyl ethan-1-ol, naproxen impurity k
PubChem CID 575523
IUPAC Name (1R)-1-(6-methoxynaphthalen-2-yl)ethan-1-ol
SMILES COC1=CC=C2C=C(C=CC2=C1)[C@@H](C)O

Specifications

Melting Point 113°C
Color White
Quantity 5 g
Formula Weight 202.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(1-Hydroxyethyl)-6-methoxynaphthalene