missing translation for 'onlineSavingsMsg'
Learn More

2-(1,3-Dioxolan-2-yl)ethyltriphenylphosphonium Bromide 97.0+%, TCI America™

Catalog No. D20565G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20565G 5 g
D205625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D20565G Supplier TCI America Supplier No. D20565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 86608-70-0
Molecular Formula C23H24BrO2P
Molecular Weight (g/mol) 443.321
MDL Number MFCD00075119
InChI Key ZCJKBPSRKLHANV-UHFFFAOYSA-M
Synonym 2-1,3-dioxolan-2-yl ethyltriphenylphosphonium bromide, 2-1,3-dioxolan-2-yl ethyl triphenylphosphonium bromide, phosphonium, 2-1,3-dioxolan-2-yl ethyl triphenyl-, bromide, 2-1,3-dioxolan-2-yl ethyl triphenylphosphanium bromide, acmc-1bl1m, 2-1,3-dioxolan-2-yl ethyltrip, phosphonium, 2-1,3-dioxolan-2-yl ethyl triphenyl-, bromide 1:1, 2-1,3-dioxolan-2-yl ethyltriphenylphosp, 2-1,3-dioxolan-2-yl ethyltriphenylphosphoniumbromide
PubChem CID 2733834
IUPAC Name 2-(1,3-dioxolan-2-yl)ethyl-triphenylphosphanium;bromide
SMILES C1COC(O1)CC[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[Br-]

Specifications

Melting Point 155°C
Quantity 5 g
Formula Weight 443.32
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(1,3-Dioxolan-2-yl)ethyltriphenylphosphonium Bromide