missing translation for 'onlineSavingsMsg'
Learn More

2-(1,3-Dioxolan-2-yl)ethylboronic acid pinacol ester, 97%

Catalog No. AAH2607103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2607103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2607103 Supplier Thermo Scientific Chemicals Supplier No. H2607103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1073354-07-0 | C11H21BO4 | 228.095 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1073354-07-0
Molecular Formula C11H21BO4
Molecular Weight (g/mol) 228.095
MDL Number MFCD03788722
InChI Key DNBRLKJRBDIKOO-UHFFFAOYSA-N
Synonym 2-2-1,3-dioxolan-2-yl ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-1,3-dioxolan-2-yl ethylboronic acid pinacol ester, 2-1,3-dioxolan-2-yl-1-ethylboronic acid pinacol ester
PubChem CID 46739008
IUPAC Name 2-[2-(1,3-dioxolan-2-yl)ethyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CCC2OCCO2

Specifications

Boiling Point 100°C (1.5 mmHg)
Quantity 1 g
Formula Weight 228.1
Percent Purity 97%
Chemical Name or Material 2-(1,3-Dioxolan-2-yl)ethylboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only