missing translation for 'onlineSavingsMsg'
Learn More

(1S)-(-)-alpha-Pinene, 98%

Catalog No. p-4509873
Change view
Click to view available options
Quantity:
5 mL
250 mL
500 mL
2.5 L
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC131270050 5 mL Glass bottle
AC131272500 250 mL Glass bottle
AC131275000 500 mL Glass bottle
AC131270025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC131270050 Supplier Thermo Scientific Chemicals Supplier No. 131270050
Only null left

CAS: 7785-26-4 | C10H16 | 136.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7785-26-4
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
InChI Key GRWFGVWFFZKLTI-IUCAKERBSA-N
Synonym --alpha-pinene, 1s,5s-2,6,6-trimethylbicyclo 3.1.1 hept-2-ene, 1s---alpha-pinene, unii-tzr3gm95pr, dl-alpha-pinene, alpha-pinene,-, 1s,5s-alpha-pinene, tzr3gm95pr, 1s,5s-2-pinene, bicyclo 3.1.1 hept-2-ene, 2,6,6-trimethyl-, 1s,5s
PubChem CID 440968
ChEBI CHEBI:28660
IUPAC Name (1S,5S)-4,6,6-trimethylbicyclo[3.1.1]hept-3-ene
SMILES CC1=CCC2CC1C2(C)C

Specifications

Melting Point -64.0°C
Color Colorless
Density 0.8595g/mL
Boiling Point 155.0°C to 156.0°C
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4650 to 1.4670
Quantity 5 mL
Beilstein 05,146
Specific Gravity 0.8595
Merck Index 14,7445
Specific Rotation Condition - 41.50 (20.00°C c=neat)
Specific Rotation − 41.50
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform,alcohol and ether
Formula Weight 136.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (1S)-(-)-α-Pinene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Harmful if swallowed.
May cause an allergic skin reaction.
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
If skin irritation or rash occurs: Get medical advice/attention.
IF ON SKIN (or hair): Take off immediately all contam
EINECSNumber 232-077-3

RUO – Research Use Only