missing translation for 'onlineSavingsMsg'
Learn More

(1S,4S)-(-)-2-Boc-2,5-diazabicyclo[2.2.1]heptane, 97%

Catalog No. AAL2013106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2013106 5 g
AAL2013103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2013106 Supplier Thermo Scientific Chemicals Supplier No. L2013106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 113451-59-5 | C10H18N2O2 | 198.266 g/mol

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Air sensitive. Store away from air, carbon dioxide, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep it refrigerated.

Chemical Identifiers

CAS 113451-59-5
Molecular Formula C10H18N2O2
Molecular Weight (g/mol) 198.266
MDL Number MFCD01569250
InChI Key UXAWXZDXVOYLII-YUMQZZPRSA-N
Synonym 1s,4s-2-boc-2,5-diazabicyclo 2.2.1 heptane, tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylate, 1s,4s-tert-butyl 2,5-diazabicyclo 2.2.1 heptane-2-carboxylate, 1s,4s-n-tert-butoxycarbonyl-2,5-diazabicyclo 2.2.1 heptane, 1s,4s---2-boc-2,5-diazabicyclo 2.2.1 heptane, tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptan-2-carboxylate, 1s,4s-2,5-diaza-bicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester, 1s,4s-n-boc-2,5-diazabicyclo 2.2.1 heptane, 1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester, tert-butyl 1s,4s---2,5-diazabicyclo 2.2.1 heptane-2-carboxylate
PubChem CID 11521263
IUPAC Name tert-butyl (1S,4S)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate
SMILES CC(C)(C)OC(=O)N1CC2CC1CN2

Specifications

Melting Point 74°C to 76°C
Quantity 5 g
Beilstein 6460311
Sensitivity Air sensitive
Solubility Information Insoluble in water.
Optical Rotation −44° (c=1 in Chloroform)
Formula Weight 198.26
Percent Purity 97%
Chemical Name or Material (1S,4S)-(-)-2-Boc-2,5-diazabicyclo[2.2.1]heptane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only