missing translation for 'onlineSavingsMsg'
Learn More

(1S,4S)-(-)-2-BOC-2,5-diazabicyclo[2.2.1]heptane, 95%

Catalog No. AC398040050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC398040050 5 g Glass bottle
AC398040010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC398040050 Supplier Thermo Scientific Chemicals Supplier No. 398040050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 113451-59-5 | C10H18N2O2 | 198.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 113451-59-5
Molecular Formula C10H18N2O2
Molecular Weight (g/mol) 198.26
MDL Number MFCD01569250
InChI Key UXAWXZDXVOYLII-YUMQZZPRSA-N
Synonym 1s,4s-2-boc-2,5-diazabicyclo 2.2.1 heptane, tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylate, 1s,4s-tert-butyl 2,5-diazabicyclo 2.2.1 heptane-2-carboxylate, 1s,4s-n-tert-butoxycarbonyl-2,5-diazabicyclo 2.2.1 heptane, 1s,4s---2-boc-2,5-diazabicyclo 2.2.1 heptane, tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptan-2-carboxylate, 1s,4s-2,5-diaza-bicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester, 1s,4s-n-boc-2,5-diazabicyclo 2.2.1 heptane, 1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester, tert-butyl 1s,4s---2,5-diazabicyclo 2.2.1 heptane-2-carboxylate
PubChem CID 11521263
IUPAC Name tert-butyl (1S,4S)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate
SMILES CC(C)(C)OC(=O)N1CC2CC1CN2

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 74.0°C to 76.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Rotation -41° to -47° (20°C, 589nm) (C=1, chloroform)
Solubility Information Solubility in water: insoluble
Formula Weight 198.26
Percent Purity 95%
Physical Form Powder
Chemical Name or Material (1S,4S)-(-)-2-BOC-2,5-diazabicyclo[2.2.1]heptane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only