missing translation for 'onlineSavingsMsg'
Learn More

(1S,4R)-1,2,3,4-Tetrahydro-1,4-epoxynaphthalene-1-carboxylic Acid 97.0+%, TCI America™

Catalog No. T3002100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3002100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3002100MG Supplier TCI America Supplier No. T3002100MG

Chemical Identifiers

CAS 1244954-13-9
Molecular Formula C11H10O3
Molecular Weight (g/mol) 190.198
InChI Key ZRBQIRRMWRKZTO-KOLCDFICSA-N
Synonym (S)-THENA
PubChem CID 91972069
SMILES C1CC2(C3=CC=CC=C3C1O2)C(=O)O

Specifications

Melting Point 138°C
Color White
Quantity 100 mg
Formula Weight 190.20
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (1S,4R)-1,2,3,4-Tetrahydro-1,4-epoxynaphthalene-1-carboxylic Acid