missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S,3R,5S)-2,3-Pinanediol, 99%

Catalog No. AAL1405314
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1405314 25 g
AAL1405303 1 g
AAL1405306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1405314 Supplier Thermo Scientific Chemicals Supplier No. L1405314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18680-27-8 | C10H18O2 | 170.252 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18680-27-8
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.252
MDL Number MFCD00077851
InChI Key MOILFCKRQFQVFS-OORONAJNSA-N
Synonym 1s,2s,3r,5s-+-2,3-pinanediol, 1s,2s,3r,5s-2,6,6-trimethylbicyclo 3.1.1 heptane-2,3-diol, 1s,2s,3r,5s-+-pinanediol, +-pinanediol, pinanediol, +, unii-y6zcv4avra, +-2-hydroxyisopinocampheol, y6zcv4avra, 1s,2s,3r,5s-2,3-pinanediol, bicyclo 3.1.1 heptane-2,3-diol, 2,6,6-trimethyl-, 1s,2s,3r,5s
PubChem CID 10219606
IUPAC Name (1S,3R,4S,5S)-4,6,6-trimethylbicyclo[3.1.1]heptane-3,4-diol
SMILES CC1(C2CC1C(C(C2)O)(C)O)C

Specifications

Melting Point 56°C to 58°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 1853591
Optical Rotation +8.7° (c=6.5 in Toluene)
Formula Weight 170.25
Percent Purity 99%
Chemical Name or Material (1S,2S,3R,5S)-2,3-Pinanediol
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only