missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-(+)-1,2-Diaminocyclohexane, 98%, Thermo Scientific™

Catalog No. AC310220050
Change view
Click to view available options
Glass bottle; 5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC310220050 Glass bottle; 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC310220050 Supplier Thermo Scientific Chemicals Supplier No. 310220050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 21436-03-3
CAS Min % 97.5
CAS Max % 100.0
Boiling Point 104.0°C to 110.0°C (40.0 mmHg)
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H14N2
Linear Formula C6H10(NH2)2
MDL Number MFCD00062986,MFCD00063747
Quantity 5g
Specific Rotation Condition +25.25 (20.00°C 589nm,c=5,1 M HCl)
Synonym 1s,2s-+-1,2-diaminocyclohexane, 1s,2s-cyclohexane-1,2-diamine, 1s,2s-+-1,2-cyclohexanediamine, s-dach, 1s,2s-1,2-diaminocyclohexane, 1s,2s-diaminocyclohexane, ss-diaminocyclohexane, 1s-+-trans-1,2-diaminocyclohexane, +-s,s-1,2-diaminocyclohexane, unii-d2v3lyg9f0
Specific Rotation + 25.25
Solubility Information Solubility in water: soluble in water.
InChI Key SSJXIUAHEKJCMH-UHFFFAOYNA-N
SMILES NC1CCCCC1N
IUPAC Name (1S,2S)-cyclohexane-1,2-diamine
Molecular Weight (g/mol) 114.19
PubChem CID 479307
Formula Weight 114.19
Percent Purity 98%
Chemical Name or Material (1S, 2S)-(+)-1, 2-Diaminocyclohexane, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only