missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-(+)-1,2-Diaminocyclohexane, 98%

Catalog No. AAL1407206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1407206 5 g
AAL1407203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1407206 Supplier Thermo Scientific Chemicals Supplier No. L1407206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Alfa Aesar (1S,2S)-(+)-1,2-Diaminocyclohexane, 98%, Quantity: 5g, Boiling Point: 104 deg.C to 110 deg.C (40mmHg), Melting Point: 43 deg.C to 45 deg.C, Molecular Weight: 114.192, Percent Purity: 98%, Beilstein: 2801645, CAS: 21436-03-3, DOT Information: Hazard Class: 8; Packaging Group: II | CAS: 21436-03-3 | C6H14N2 | 114.19 g/mol

For synthesis of optically active products

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21436-03-3
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.19
MDL Number MFCD00062986,MFCD00063747
InChI Key SSJXIUAHEKJCMH-UHFFFAOYNA-N
Synonym 1s,2s-+-1,2-diaminocyclohexane, 1s,2s-cyclohexane-1,2-diamine, 1s,2s-+-1,2-cyclohexanediamine, s-dach, 1s,2s-1,2-diaminocyclohexane, 1s,2s-diaminocyclohexane, ss-diaminocyclohexane, 1s-+-trans-1,2-diaminocyclohexane, +-s,s-1,2-diaminocyclohexane, unii-d2v3lyg9f0
PubChem CID 479307
SMILES NC1CCCCC1N

Specifications

Melting Point 43°C to 45°C
Boiling Point 104°C to 110°C (40 mmHg)
Flash Point 70°C (158°F)
Quantity 5 g
UN Number UN3259
Beilstein 2801645
Sensitivity Air sensitive
Optical Rotation +25.5° (c=5 in 1N HCl)
IUPAC Name cyclohexane-1,2-diamine
Formula Weight 114.19
Percent Purity 98%
Chemical Name or Material (1S,2S)-(+)-1,2-Diaminocyclohexane
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only