missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-1,2-Cyclohexanedicarboxylic Acid 98.0+%, TCI America™

Catalog No. C19545G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C19545G 5 g
C19541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C19545G Supplier TCI America Supplier No. C19545G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 21963-41-7
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00151258,MFCD00001464
InChI Key QSAWQNUELGIYBC-UHFFFAOYNA-N
Synonym 1s,2s-cyclohexane-1,2-dicarboxylic acid, 1s,2s-1,2-cyclohexanedicarboxylic acid, 1,2-cyclohexanedicarboxylic acid, trans, 1,2-cyclohexanedicarboxylic acid, 1s,2s, 1,2-cyclohexanedicarboxylicacid, 1s,2s, pubchem19269, trans-hexahydrophthalic acid, 1alpha,2beta-cyclohexanedicarboxylic acid, 1s,2s-1,2-cyclohexanedicarboxylicacid
PubChem CID 75315
IUPAC Name cyclohexane-1,2-dicarboxylic acid
SMILES OC(=O)C1CCCCC1C(O)=O

Specifications

Melting Point 184°C
Color White
Quantity 5 g
Formula Weight 172.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (1S,2S)-1,2-Cyclohexanedicarboxylic Acid
EINECSNumber (3)-2678
TSCA No