missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-(+)-1,2-Cyclohexanediamine 98.0+%, TCI America™

Catalog No. C14485G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14485G 5 g
C144825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14485G Supplier TCI America Supplier No. C14485G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 21436-03-3
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.19
MDL Number MFCD00062986,MFCD00063747
InChI Key SSJXIUAHEKJCMH-UHFFFAOYNA-N
Synonym 1s,2s-+-1,2-diaminocyclohexane, 1s,2s-cyclohexane-1,2-diamine, 1s,2s-+-1,2-cyclohexanediamine, s-dach, 1s,2s-1,2-diaminocyclohexane, 1s,2s-diaminocyclohexane, ss-diaminocyclohexane, 1s-+-trans-1,2-diaminocyclohexane, +-s,s-1,2-diaminocyclohexane, unii-d2v3lyg9f0
PubChem CID 479307
IUPAC Name cyclohexane-1,2-diamine
SMILES NC1CCCCC1N

Specifications

Melting Point 43°C
Color White-Yellow
Boiling Point 81°C
Quantity 5 g
UN Number 3259
Formula Weight 114.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material (1S,2S)-(+)-1,2-Cyclohexanediamine
EINECSNumber (3)-4230
TSCA No