missing translation for 'onlineSavingsMsg'
Learn More

(1S,2R,6S,7R)-4,4-Dimethyl-3,5-dioxa-8-azatricyclo[5.2.1.0(2,6)]decan-9-one, 95%, 98% ee

Catalog No. AC362200010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC362200010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC362200010 Supplier Thermo Scientific Chemicals Supplier No. 362200010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 178032-63-8 | C9H13NO3 | 183.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 178032-63-8
Molecular Formula C9H13NO3
Molecular Weight (g/mol) 183.2
MDL Number MFCD01320865
InChI Key CSGIROCFQXHTKS-WNJXEPBRSA-N
Synonym 1s,2r,6s,7r-4,4-dimethyl-3,5-dioxa-8-azatricyclo 5.2.1.0 2,6 decan-9-one, 3as,4r,7s,7ar-2,2-dimethyltetrahydro-4,7-methano 1,3 dioxolo 4,5-c pyridin-6 3ah-one, 2s,7s,1r,6r-4,4-dimethyl-3,5-dioxa-9-azatricyclo 5.2.1.0<2,6> decan-8-one, 1s,2r,6s,7r-4,4-dimethyl-3,5-dioxa-8-azatricyclo 5.2.1.0 2 ,? decan-9-one, 4,7-methano-1,3-dioxolo 4,5-c pyridin-6 3ah-one,tetrahydro-2,2-dimethyl-, 3as,4r,7s,7ar
PubChem CID 10219706
SMILES CC1(OC2C3CC(C2O1)NC3=O)C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 150°C to 152°C
Color White
Enantiomeric Excess 97.5% min. (GC)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 183.2
Percent Purity 95%,98% e.e
Physical Form Needle-like Crystals or Crystalline Powder
Chemical Name or Material (1S,2R,6S,7R)-4,4-Dimethyl-3,5-dioxa-8-azatricyclo[5.2.1.0(2,6)]decan-9-one, 95%, (98% e.e.)
Show More Show Less
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only