missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1S,2R)-2-(Dibutylamino)-1-phenyl-1-propanol 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 170°C |
| Quantity | 5 g |
| Formula Weight | 263.43 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | (1S,2R)-2-(Dibutylamino)-1-phenyl-1-propanol |
Safety and Handling
| TSCA | No |