missing translation for 'onlineSavingsMsg'
Learn More

(1S,2R)-1-Phenyl-2-(1-pyrrolidinyl)propan-1-ol 98.0+%, TCI America™

Catalog No. P13751G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13751G 1 g
P13755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13751G Supplier TCI America Supplier No. P13751G
Only null left

Chemical Identifiers

CAS 123620-80-4
Molecular Formula C13H19NO
Molecular Weight (g/mol) 205.30
MDL Number MFCD02093492
InChI Key FZVHJGJBJLFWEX-UHFFFAOYNA-N
Synonym 1s,2r-1-phenyl-2-pyrrolidin-1-yl propan-1-ol, 1s,2r-1-phenyl-2-1-pyrrolidinyl propan-1-ol, 1s,2r-1-phenyl-2-pyrrolidin-1-yl-propan-1-ol, 1s,2r-1-phenyl-2-1-pyrrolidinyl-1-propanol, 1s,2r-n,n-tetramethylenenorephedrine, 1-phenyl-2-pyrrolidin-1-yl propan-1-ol, 1s,2r, 1-pyrrolidineethanol, beta-methyl-alpha-phenyl-, alphas,betar, pubchem8276, pubchem14465, 1s,2r-1-phenyl-2-pyrrolizino-1-propanol
PubChem CID 9942483
IUPAC Name 1-phenyl-2-(pyrrolidin-1-yl)propan-1-ol
SMILES CC(C(O)C1=CC=CC=C1)N1CCCC1

Specifications

Melting Point 48°C
Color White-Yellow
Quantity 1 g
Formula Weight 205.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (1S,2R)-1-Phenyl-2-(1-pyrrolidinyl)propan-1-ol