missing translation for 'onlineSavingsMsg'
Learn More

(1R)-(-)-Menthyl acetate, 98%

Catalog No. AAL14095MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL14095MD 250 mg
AAL1409503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL14095MD Supplier Thermo Scientific Chemicals Supplier No. L14095MD
Only null left

CAS: 2623-23-6 | C12H22O2 | 198.306 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2623-23-6
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.306
MDL Number MFCD00001482
InChI Key XHXUANMFYXWVNG-OIKLOGQESA-N
Synonym 1r---menthyl acetate, 5r-2-isopropyl-5-methylcyclohexyl acetate, --menthylacetate, --1r-menthyl acetate
PubChem CID 12732529
IUPAC Name [(5R)-5-methyl-2-propan-2-ylcyclohexyl] acetate
SMILES CC1CCC(C(C1)OC(=O)C)C(C)C

Specifications

Density 0.925
Boiling Point 229°C to 231°C
Flash Point 77°C (170°F)
Refractive Index 1.447
Quantity 250 mg
Beilstein 2208505
Merck Index 14,5839
Optical Rotation −80.5° (c=8 in Benzene)
Formula Weight 198.31
Percent Purity 98%
Chemical Name or Material (1R)-(-)-Menthyl acetate
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 220-076-0
RTECSNumber AI5900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only