missing translation for 'onlineSavingsMsg'
Learn More

(1R)-(+)-alpha-Pinène 97,0+%, TCI America™

Numéro de catalogue. P109925ML
Modifier l'affichage
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
P109925ML 25 mL
P1099500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. P109925ML Fournisseur TCI America Code fournisseur P109925ML

Identifiants chimiques

CAS 7785-70-8
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00001346
InChI Key GRWFGVWFFZKLTI-RKDXNWHRSA-N
Synonym +-alpha-pinene, 1r,5r-2,6,6-trimethylbicyclo 3.1.1 hept-2-ene, 1r-+-alpha-pinene, 1r,5r-alpha-pinene, unii-h6cm4twh1w, +-pinene, h6cm4twh1w, 1r,5r-2-pinene, bicyclo 3.1.1 hept-2-ene, 2,6,6-trimethyl-, 1r,5r, +-pin-2 3-ene
PubChem CID 82227
ChEBI CHEBI:28261
IUPAC Name (1R,5R)-2,6,6-triméthylbicyclo[3.1.1]hept-2-ène
SMILES CC1=CC[C@@H]2C[C@H]1C2(C)C

Spécifications

Color Incolore
Boiling Point 156°C
Quantity 25 mL
UN Number 2368
Formula Weight 136.24
Percent Purity ≥97.0% (GC)
Physical Form Liquide
Chemical Name or Material (1R)-(+)-alpha-Pinene
EINECSNumber (4)-0593&(8)-0497
TSCA Yes