missing translation for 'onlineSavingsMsg'
Learn More

(1R,2S,5R)-(-)-Menthyl (S)-p-Toluenesulfinate 98.0+%, TCI America™

Catalog No. M10445G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10445G 5 g
M104425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M10445G Supplier TCI America Supplier No. M10445G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1517-82-4
Molecular Formula C17H26O2S
Molecular Weight (g/mol) 294.453
MDL Number MFCD00010192
InChI Key NQICGNSARVCSGJ-DSRNLFJRSA-N
Synonym 1r,2s,5r---menthyl s-p-toluenesulfinate, 4-methylbenzenesulfinic acid 1r,5r-2-isopropyl-5-methylcyclohexyl ester
PubChem CID 101348873
IUPAC Name [(1R,5R)-5-methyl-2-propan-2-ylcyclohexyl] 4-methylbenzenesulfinate
SMILES CC1CCC(C(C1)OS(=O)C2=CC=C(C=C2)C)C(C)C

Specifications

Melting Point 105°C
Color White-Yellow
Quantity 5 g
Formula Weight 294.45
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (1R,2S,5R)-(-)-Menthyl (S)-p-Toluenesulfinate
TSCA No
Recommended Storage Freezer