missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-Bis[(2-methoxyphenyl)phenylphosphino]ethane, 90%, Thermo Scientific™

Identifiants chimiques

CAS 55739-58-7
Molecular Formula C28H28O2P2
Molecular Weight (g/mol) 458.48
MDL Number MFCD05863546
InChI Key QKZWXPLBVCKXNQ-UHFFFAOYNA-N
Synonym r,r-dipamp, 1,2-bis r-2-methoxyphenyl phenyl phosphino ethane, unii-blj831owlw, --dipamp, r,r-ethylenebis 2-methoxyphenyl phenylphosphine, r,r-1,2-bis 2-methoxyphenyl phenylphosphino ethane, blj831owlw, s,s-dipamp, 1r,2r---bis 2-methoxyphenyl phenylphosphino ethane
PubChem CID 10884975
IUPAC Name (R)-(2-methoxyphenyl)-[2-[(2-methoxyphenyl)-phenylphosphanyl]ethyl]-phenylphosphane
SMILES COC1=CC=CC=C1P(CCP(C1=CC=CC=C1)C1=CC=CC=C1OC)C1=CC=CC=C1
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantity & Availability  
AC379352500
Afficher les documents Détails de la promotion
N/A 250mg Glass bottle N/A
N/A
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Description

Description

Spécifications

Identifiants chimiques

55739-58-7
458.48
QKZWXPLBVCKXNQ-UHFFFAOYNA-N
10884975
COC1=CC=CC=C1P(CCP(C1=CC=CC=C1)C1=CC=CC=C1OC)C1=CC=CC=C1
C28H28O2P2
MFCD05863546
r,r-dipamp, 1,2-bis r-2-methoxyphenyl phenyl phosphino ethane, unii-blj831owlw, --dipamp, r,r-ethylenebis 2-methoxyphenyl phenylphosphine, r,r-1,2-bis 2-methoxyphenyl phenylphosphino ethane, blj831owlw, s,s-dipamp, 1r,2r---bis 2-methoxyphenyl phenylphosphino ethane
(R)-(2-methoxyphenyl)-[2-[(2-methoxyphenyl)-phenylphosphanyl]ethyl]-phenylphosphane

Spécifications

55739-58-7
100.0
90%
MFCD05863546
-80° to -85° (20°C, 589nm) (C=1, CHCl3)
QKZWXPLBVCKXNQ-UHFFFAOYNA-N
(R)-(2-methoxyphenyl)-[2-[(2-methoxyphenyl)-phenylphosphanyl]ethyl]-phenylphosphane
10884975
90%
88.0
Authentic
C28H28O2P2
15, 3333
r,r-dipamp, 1,2-bis r-2-methoxyphenyl phenyl phosphino ethane, unii-blj831owlw, --dipamp, r,r-ethylenebis 2-methoxyphenyl phenylphosphine, r,r-1,2-bis 2-methoxyphenyl phenylphosphino ethane, blj831owlw, s,s-dipamp, 1r,2r---bis 2-methoxyphenyl phenylphosphino ethane
COC1=CC=CC=C1P(CCP(C1=CC=CC=C1)C1=CC=CC=C1OC)C1=CC=CC=C1
458.48
458.48
(1R,2R)-Bis[(2-methoxyphenyl)phenylphosphino]ethane
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

FDS
Documents

Documents

RUO – Research Use Only