missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 1M TEAB and 50% Hydroxylamine Buffers for TMT Experiments
Description
Pre-formulated TEAB and 50% hydroxylamine designed for use in TMT labeling and quenching of peptides.
Triethylamonium bicarbonate (TEAB) is a 10X dissolution buffer for isobaric mass tag labeling experiments. 50% hydroxylamine is a 10X quenching reagent for amine-reactive labeling for TMT experiments.
Spécifications
Spécifications
| Product Type | TEAB |
| Product Line | TMT |
| Content And Storage | Store protected from direct sunlight in a dry, cool, and well-ventilated area. |
| Starting Material | Cell Lysate |
| Workflow Step | Protein Labeling |
| Quantity | 50 mL |