missing translation for 'onlineSavingsMsg'
Learn More

1H-Undecafluoropentane 97.0+%, TCI America™

Catalog No. U00765G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. U00765G Supplier TCI America Supplier No. U00765G
Only null left

Chemical Identifiers

CAS 375-61-1
Molecular Formula C5HF11
Molecular Weight (g/mol) 270.045
MDL Number MFCD08458172
InChI Key WXFBZGUXZMEPIR-UHFFFAOYSA-N
Synonym 1H-Perfluoropentane
PubChem CID 9816874
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5-undecafluoropentane
SMILES C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Color Colorless
Quantity 5 g
Physical Form Clear Liquid at 20°C
Boiling Point 46°C
Formula Weight 270.05
Percent Purity ≥97.0% (GC)
Chemical Name or Material 1H-Undecafluoropentane