missing translation for 'onlineSavingsMsg'
Learn More

1H,1H-Undecafluoro-1-hexanol 96.0+%, TCI America™

Catalog No. U00745G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. U00745G Supplier TCI America Supplier No. U00745G
Only null left

Chemical Identifiers

CAS 423-46-1
Molecular Formula C6H3F11O
Molecular Weight (g/mol) 300.07
MDL Number MFCD04038338
InChI Key QZFZPVVDBGXQTB-UHFFFAOYSA-N
Synonym 1H,1H-Perfluoro-1-hexanol, 1,1-Dihydroperfluorohexanol
PubChem CID 2782405
IUPAC Name 2,2,3,3,4,4,5,5,6,6,6-undecafluorohexan-1-ol
SMILES OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Color Colorless
Boiling Point 128°C
Quantity 5 g
Formula Weight 300.07
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1H,1H-Undecafluoro-1-hexanol