missing translation for 'onlineSavingsMsg'
Learn More

1H,1H-Tricosafluoro-1-dodecanol 98.0+%, TCI America™

Catalog No. T33811G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33811G 1 g
T33815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33811G Supplier TCI America Supplier No. T33811G

Chemical Identifiers

CAS 423-65-4
Molecular Formula C12H3F23O
Molecular Weight (g/mol) 600.12
MDL Number MFCD00153235
InChI Key SHTZQFTXUMCALC-UHFFFAOYSA-N
Synonym 1h,1h-perfluoro-1-dodecanol, 1h,1h-perfluorododecan-1-ol, 1h,1h-perfluorododecanol, tricosafluoroundecylmethanol, 1h,1h-tricosafluoro-1-dodecanol, 1h,1h-perfluoro-1-lauryl alcohol
PubChem CID 2760321
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecan-1-ol
SMILES OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Melting Point 119°C
Color White
Boiling Point 224°C
Quantity 1 g
Formula Weight 600.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1H,1H-Tricosafluoro-1-dodecanol