missing translation for 'onlineSavingsMsg'
Learn More

1H,1H-Nonadecafluoro-1-decanol 98.0+%, TCI America™

Catalog No. N08145G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N08145G Supplier TCI America Supplier No. N08145G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 307-37-9
Molecular Formula C10H3F19O
Molecular Weight (g/mol) 500.103
MDL Number MFCD00153234
InChI Key NIRPXSQCRWXHNZ-UHFFFAOYSA-N
Synonym 1h,1h-perfluoro-1-decanol, 1h,1h-nonadecafluoro-1-decanol, 1h,1h-perfluorodecan-1-ol, nonadecafluorononyl methanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecakis fluoranyl decan-1-ol
PubChem CID 2733275
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluorodecan-1-ol
SMILES C(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

Melting Point 88°C
Color White
Boiling Point 136°C
Quantity 5 g
Formula Weight 500.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1H,1H-Nonadecafluoro-1-decanol