missing translation for 'onlineSavingsMsg'
Learn More

1H-1,2,4-Triazole-1-carboxamidine hydrochloride, 98%

Catalog No. AAH6047514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6047514 25 g
AAH6047506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6047514 Supplier Thermo Scientific Chemicals Supplier No. H6047514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 19503-26-5 | C3H6ClN5 | 147.566 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19503-26-5
Molecular Formula C3H6ClN5
Molecular Weight (g/mol) 147.566
MDL Number MFCD03095468
InChI Key JDDXNENZFOOLTP-UHFFFAOYSA-N
Synonym 1h-1,2,4-triazole-1-carboximidamide hydrochloride, 1h-1,2,4-triazole-1-carboxamidine monohydrochloride, 1h-1,2,4-triazole-1-carboxamidine hydrochloride, 1,2,4-triazole-1-carboximidamide hydrochloride, 1h-1,2,4-triazole-1-carboximidamide, monohydrochloride, c3h5n5.clh, ksc496s5d, 1,2,4-triazolecarboxamidine, chloride, 1-amidino-1,2,4-triazole hydrochloride, 1-amidino-1h-1,2,4-triazole hydrochloride
PubChem CID 16218515
IUPAC Name 1,2,4-triazole-1-carboximidamide;hydrochloride
SMILES C1=NN(C=N1)C(=N)N.Cl

Specifications

Melting Point 215°C to 220°C
Quantity 25 g
Beilstein 9500271
Formula Weight 147.57
Percent Purity 98%
Chemical Name or Material 1H-1,2,4-Triazole-1-carboxamidine hydrochloride
Hazard Category H302-H319
Hazard Statement GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only