Learn More
Electron Microscopy Sciences 16% Paraformaldehyde Aqueous Solution, EM Grade, Ampoule 10 ML
Supplier:  Electron Microscopy Sciences EMS 15710
Encompass
Paraformaldehyde methanol free solution. A more efficient and rapid penetrant fixative used in combination with Glutaraldehyde and Acrolein fixatives. Easy to break, prescored, 10 ml ampoules sealed under inert gas.
Grade: EM Grade
Package: Ampoule
Numéro de catalogue. 50980487