missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 1536-Well Nontreated Polystyrene Plates
GREENER_CHOICE

Catalog No. 07202511
Change view
Click to view available options
Color:
Black
Black, Clear Bottom
White
Volume (Metric) Working:
8 μL
10 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Volume (Metric) Working
07202511 White 8 μL
07202510 Black 10 μL
0720114 Black, Clear Bottom 8 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 07202511 Supplier Corning™ Supplier No. 3725
Get Up to $2,500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Superior flatness minimizes variability within a plate and between plates

  • Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
  • Flat bottom wells are compatible with automated dispensers including the PerkinElmer™ FlexDrop™
  • Working volume of up to 10 μL
Generic Bar Code
  • Ensures compatibility with most bar code scanning and software systems
  • To avoid mix-ups, the surface treatment of the microplate wells is easily identified in the human readable portion of the bar code (NT = not treated; TC = tissue culture treated; NB = nonbinding surface)
  • Labels applied to all four sides of the plate No sequence number duplicates

Order Info

Packaged in bags of 10

Specifications

Format Standard
Lid Without Lid
Array 32 x 48
Color White
For Use With (Equipment) Automated dispensers
Material Polystyrene
No. of Wells 1536
Volume (Metric) Working 8 μL
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
For Use With (Application) For barcode scanning and software systems
No. per Case 50/Cs.
No. per Pack 10/Pk.
Show More Show Less