Reaction Vessel Lid; 4-Neck; Flange I.D: 150mm; Center Neck: 29/42; Side Necks: 29/42; Fits Reaction Vessels (CG-1920 through CG-1936); Contains flat ground surface allowing it to be greased or used with an o-ring
Numéro de catalogue.
CG194105
Fournisseur
Chemglass IncCode fournisseur
CG194105
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Reaction Vessel Lid; 4-Neck; Flange I.D: 150mm; Center Neck: 29/42; Side Necks: 29/42; Fits Reaction Vessels (CG-1920 through CG-1936); Contains flat ground surface allowing it to be greased or used with an o-ring