Learn More
Saint-Gobain Bioprocess Bag, 150 mL, 2 Ports, 12 in Tubing
Numéro de catalogue. NC2230944
Encompass
Single-use bioprocess bag designed for small-volume fluid handling and transfer in bioprocessing workflows. The 150 mL 2D bag includes two ports fitted with 12 in (30 cm) tubing and is supplied 20 per box. The bag is constructed from a multilayer film optimized for bioprocess applications, providing chemical compatibility and reduced extractables for process use.
  • 150 mL capacity.
  • Standard 2D bag configuration.
  • Two ports with 12 in tubing each.
  • Multilayer film construction optimized for bioprocess use.
  • Supplied 20 bags per box.
Numéro de catalogue. NC2230944
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Single-use bioprocess bag designed for small-volume fluid handling and transfer in bioprocessing workflows. The 150 mL 2D bag includes two ports fitted with 12 in (30 cm) tubing and is supplied 20 per box. The bag is constructed from a multilayer film optimized for bioprocess applications, providing chemical compatibility and reduced extractables for process use.
  • 150 mL capacity.
  • Standard 2D bag configuration.
  • Two ports with 12 in tubing each.
  • Multilayer film construction optimized for bioprocess use.
  • Supplied 20 bags per box.