missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 150 Homogenizer Package
GREENER_CHOICE

Catalog No. 15340168
Change view
Click to view available options
Voltage:
115 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
15340168 115 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning: Stainles Steel bearings cannot be autoclaved.
Catalog No. 15340168 Supplier Fisherbrand™ Supplier No. 15340168

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Warning: Stainles Steel bearings cannot be autoclaved.

Fisher Scientific™ 150 Homogenizer Package includes everything needed to process a wide range of laboratory samples, typically in less than 30 seconds. Includes motor, stand assembly and 3 generator probes.

  • Processes laboratory samples, from 200μL to 100mL
  • Powerful 150w motor
  • Lightweight ergonomic design
  • Quiet operation, under 72db
  • Available with disposable generator probes (adapter for use is included)
  • Cross-contamination can be completely eliminated with the use of plastic generator probes
  • Includes 3 stainless steel generator probes; 5 x 75, 7 x 115, and 10 x 115 mm

Certifications

CE

Warranty

One year

Specifications

Product Type Tissue Homogenizer
Speed 5000 to 35,000 rpm
Voltage 115 V
Volume Range (Metric) 0.2 to 100 mL
Diameter (English) 2 in.
Diameter (Metric) 5.1 cm
Height (English) 6.5 in.
Height (Metric) 16 cm
For Use With (Application) Tissue homogenization, tissue disruption, protein extraction, RNA extraction, DNA extraction, metabolite extraction, emulsification, proteomics, PCR, enzyme extraction, high speed stirring, powder suspension, genomics, metabolomics
Includes 150 W motor, Flat bottom stainless steel generator probe (5 x 75 mm), Sawtooth stainless steel generator probe (7 x 115 mm), Sawtooth stainless steel generator probe (10 x 115 mm), Stand clamp, Motor stand assembly, Disposable probe
Weight (English) 16.4 oz.
Weight (Metric) 466 g
Show More Show Less