missing translation for 'onlineSavingsMsg'
Learn More

15-Pentadecanolactone 98.0+%, TCI America™

Catalog No. P098525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P098525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P098525G Supplier TCI America Supplier No. P098525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 106-02-5
Molecular Formula C15H28O2
Molecular Weight (g/mol) 240.387
MDL Number MFCD00039667
InChI Key FKUPPRZPSYCDRS-UHFFFAOYSA-N
Synonym cyclopentadecanolide, exaltolide, pentadecanolide, 15-pentadecanolide, pentalide, thibetolide, pentadecan-15-olide, pentadecalactone, muskalactone, muskolactone
PubChem CID 235414
IUPAC Name oxacyclohexadecan-2-one
SMILES C1CCCCCCCOC(=O)CCCCCC1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 240.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 15-Pentadecanolactone
EINECSNumber (2)-1089
TSCA Yes