Learn More
Brady™ SPC™ Absorbents 14 Gallon Chemical Spill Kit Refill
Catalog No. 17000910
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Capacity (English) Sorbing:
10.1 gal.
Catalog No. Capacity (English) Sorbing
17000910 10.1 gal.
1 missing translation for 'options'
Catalog No. 17000910 missing translation for 'mfr' Brady™ SPC™ Absorbents missing translation for 'supplierNo' SKH14R
Refill your spill kit's absorbents, accessories, PPE and instructional guidelines so you are prepared for when a spill occurs.
SPC™ Spill Kits from Brady™ combine absorbents, accessories, PPE and instructional guidelines so you are prepared for when a spill occurs. Improve worker productivity and efficiency by placing spill kits around your facility close to areas prone to spills.
  • Refill kit only (container SKH-14 sold separately)
  • Surfactant-treated, brightly colored absorbent picks up a wide range of aggressive chemicals while remaining chemically inert
  • Ideal for fast response to critical accidents
Specifications
Capacity (English) Reservoir 14 gal.
Capacity (English) Sorbing 10.1 gal.
Capacity (Metric) Reservoir 53 L
Capacity (Metric) Sorbing 38.2 L
Material Polypropylene
Type Spill Kit
For Use With (Application) Absorbents and Spill Control
Includes 20 Chemical Pads (15 x 19 in.), 2 Chemical Socks (3 in. x 12 ft.), 2 Disposal Bags, 1 Pair of Goggles, 1 Pair of Nitrile Gloves