Learn More
Chemglass Inc Adapter, Thermometer, Inlet, 14/20 Inner Joint, 55mm Height
Supplier:  Chemglass Inc CG103814
Encompass
Adapter, Thermometer, Inlet, 14/20 Inner Joint, 55mm Height. Adapter has a tubulation at the top for use with a CG-1040 rubber fitting. Fitting permits insertion of plain stem thermometers or bleed tubes. Supplied complete with one CG-1040 rubber fitting.
Numéro de catalogue. CG103814
Il en reste null