missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 13mm Autosampler Vial Screw Thread Caps

Catalog No. 03378316
Change view
Click to view available options
Color:
Black
Black, Blue, White
Black, Ivory, Red
Black, Red, White
Natural
Septum:
Integral Molded
Ivory PTFE/Red Rubber
None
PTFE/Silicone, Pre-slit
PTFE/Silicone/PTFE
Red PTFE/White Silicone
Material:
Phenolic
Phenolic, PTFE, Silicone
Polypropylene
Polypropylene, Polyethylene
Polypropylene, PTFE, Rubber
Polypropylene, PTFE, Silicone
Product Type:
Open Top Screw Cap
Screw Cap
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Septum Material Product Type
03378316 Black, Red, White Red PTFE/White Silicone Phenolic, PTFE, Silicone Open Top Screw Cap
0337525A Black None Polypropylene Open Top Screw Cap
03378315 Black None Phenolic Open Top Screw Cap
03377106 Black, Ivory, Red Ivory PTFE/Red Rubber Polypropylene, PTFE, Rubber Open Top Screw Cap
03377108 Black, Red, White PTFE/Silicone/PTFE Polypropylene, PTFE, Silicone Open Top Screw Cap
03376250 Black, Blue, White PTFE/Silicone, Pre-slit Polypropylene, PTFE, Silicone Open Top Screw Cap
0337525C Natural Integral Molded Polypropylene, Polyethylene Screw Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 03378316 Supplier Thermo Scientific™ Supplier No. C401566A

13mm Screw Thread Caps feature a 13-425 thread finish. Available in an assortment of styles, pre-assembled caps and septa minimize contamination.

Thermo Scientific™ 13mm Screw Thread Caps feature a 13-425 thread finish and are available in an assortment of styles to match your HPLC, GC or IC application needs. Convenient pre-assembled caps and septa minimize contamination from handling. Choose from Thermo Scientific™, Thermo Scientific™ National™, and Thermo Scientific™ Chromacol™ brands.

Autosampler Vial Screw Thread Caps

Specifications

Color (Seal) Red/White
Description Phenolic, Black; Red PTFE/White Silicone septum
Color Black, Red, White
Material Phenolic, PTFE, Silicone
Septum Red PTFE/White Silicone
Quantity 100
Thickness (Metric) 1.5 mm
Closure Size 13 mm
For Use With (Equipment) 13-425 Screw Thread Vials
Product Type Open Top Screw Cap

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.