missing translation for 'onlineSavingsMsg'
Learn More

13-Azido-2,5,8,11-tetraoxatridecane 98.0+%, TCI America™

Catalog No. A272825MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A272825MG 25 mg
A2728100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A272825MG Supplier TCI America Supplier No. A272825MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 606130-90-9
Molecular Formula C9H19N3O4
Molecular Weight (g/mol) 233.268
MDL Number MFCD13184960
InChI Key FFOZZVDSANUDAE-UHFFFAOYSA-N
PubChem CID 51340962
IUPAC Name 1-azido-2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethane
SMILES COCCOCCOCCOCCN=[N+]=[N-]

Specifications

Color Yellow
Quantity 25 mg
Synonym O-(2-Azidoethyl)-O′C-methyltriethylene Glycol, mPEG4-Azide, mPEG4-N3, Methyl-PEG4-Azide
Formula Weight 233.27
Percent Purity ≥98.0% (HPLC)
Physical Form Liquid
Chemical Name or Material 13-Azido-2,5,8,11-tetraoxatridecane
TSCA No
Recommended Storage Refrigerator