missing translation for 'onlineSavingsMsg'
Learn More

(11S,12S)-9,10-Dihydro-9,10-ethanoanthracene-11,12-diamine 98.0+%, TCI America™

Catalog No. D34451G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34451G 1 g
D3445100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34451G Supplier TCI America Supplier No. D34451G
Only null left

Chemical Identifiers

CAS 138517-66-5
Molecular Formula C16H16N2
Molecular Weight (g/mol) 236.318
MDL Number MFCD06654768
InChI Key NWDYSRZJOLDMRE-CKUJCDMFSA-N
PubChem CID 11499644
SMILES C1=CC=C2C3C(C(C(C2=C1)C4=CC=CC=C34)N)N

Specifications

Melting Point 158°C
Color White-Yellow
Quantity 1 g
Formula Weight 236.32
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (11S,12S)-9,10-Dihydro-9,10-ethanoanthracene-11,12-diamine