missing translation for 'onlineSavingsMsg'
Learn More

11-Azido-3,6,9-trioxaundecanoic Acid 97.0+%, TCI America™

Catalog No. A22931G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A22931G Supplier TCI America Supplier No. A22931G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 172531-37-2
Molecular Formula C8H15N3O5
Molecular Weight (g/mol) 233.224
InChI Key GIXBCECBLAEYKA-UHFFFAOYSA-N
Synonym Azido-PEG3-acetic Acid, 2-[2-[2-(2-Azidoethoxy)ethoxy]ethoxy]acetic Acid
PubChem CID 10911448
IUPAC Name 2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]acetic acid
SMILES C(COCCOCCOCC(=O)O)N=[N+]=[N-]

Specifications

Quantity 1 g
Formula Weight 233.22
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 11-Azido-3,6,9-trioxaundecanoic Acid